Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Contactin-2/TAG1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 9 publications
Supplier: R&D Systems AF4439
This item is not returnable.
View return policy
Description
Contactin-2/TAG1 Polyclonal specifically detects Contactin-2/TAG1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.Specifications
Contactin-2/TAG1 | |
Polyclonal | |
Unconjugated | |
Q61330 | |
CNTN2 | |
Mouse myeloma cell line NS0-derived recombinant mouse Contactin-2/TAG1 Gln31-Ser1014 Accession # Q61330 | |
100 μg | |
Primary | |
Detects mouse and rat Contactin-2 in direct ELISAs. Detects human, mouse, and rat Contactin-2 in Western blots. | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Axonal glycoprotein TAG-1, Axonin-1, axonin-1 cell adhesion molecule, AXT, CNTN2, contactin 2 (axonal), contactin 2 (transiently expressed), contactin-2, Contactin2, DKFZp781D102, FLJ37193, MGC157722, TAG1, TAG-1, TAX, TAX1, TAX-1, TAX1FLJ42746, Transient axonal glycoprotein 1, transiently-expressed axonal glycoprotein | |
Goat | |
Affinity Purified | |
RUO | |
6900 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction