Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat CSRP1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF5739SP
This item is not returnable.
View return policy
Description
CSRP1 Polyclonal specifically detects CSRP1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
CSRP1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CRP, CRP1, CSRPCYRP, CYRP, cysteine and glycine-rich protein 1, Cysteine-rich protein 1, D1S181E, DKFZp686M148, LIM-domain protein | |
Goat | |
Affinity Purified | |
RUO | |
1465 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.5 ug/mL | |
P21291 | |
CSRP1 | |
E. coli-derived recombinant human CSRP1 Met1-Glu193 Accession # P21291 | |
25 μg | |
Primary | |
Detects human, mouse, and rat CSRP1 in Western blots. | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction