Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat CUGBP1/CELF1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $410.00
Specifications
Antigen | CUGBP1/CELF1 |
---|---|
Clone | 850717 |
Dilution | Western Blot 0.5 ug/mL, Immunohistochemistry 8-25 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9388SP
![]() |
R&D Systems
MAB9388SP |
25 μg |
Each for $200.50
|
|
|||||
MAB9388100
![]() |
R&D Systems
MAB9388100 |
100 μg |
Each for $410.00
|
|
|||||
Description
CUGBP1/CELF1 Monoclonal specifically detects CUGBP1/CELF1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
CUGBP1/CELF1 | |
Western Blot 0.5 ug/mL, Immunohistochemistry 8-25 ug/mL | |
Monoclonal | |
Mouse | |
Human, Mouse, Rat | |
Q92879 | |
10658 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
850717 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
BRUNOL2, BRUNOL250 kDa nuclear polyadenylated RNA-binding protein, bruno-like 2, Bruno-like protein 2, CELF1, CUG RNA-binding protein, CUG triplet repeat, RNA binding protein 1, CUG triplet repeat, RNA-binding protein 1, CUG-BP, CUG-BP- and ETR-3-like factor 1, CUGBP Elav-like family member 1, CUGBP, Elav-like family member 1, CUG-BP1, CUGBP1CUG triplet repeat RNA-binding protein 1, CUGBPCELF-1, D2Wsu101e, Deadenylation factor CUG-BP, EDEN-BP, EDEN-BP homolog, embryo deadenylation element binding protein, Embryo deadenylation element-binding protein homolog, hNab50, NAB50, NAB50CUG-BP1, NAPOR, nuclear polyadenylated RNA-binding protein, 50-kD, RNA-binding protein BRUNOL-2 | |
E. coli-derived recombinant human CUGBP1/CELF1 Met1-Gly60 Accession # Q92879 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human CUGBP1/CELF1 in direct ELISAs and Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title