Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Cyclophilin B Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5410SP
This item is not returnable.
View return policy
Description
Cyclophilin B Monoclonal specifically detects Cyclophilin B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry, Knockout Validated.Specifications
Cyclophilin B | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL, Knockout Validated | |
P23284 | |
PPIB | |
E. coli-derived recombinant human Cyclophilin B Asp34-Glu216 Accession # P23284 | |
25 μg | |
Primary | |
Detects human, mouse, and rat Cyclophilin B in Western blots. In Western blots, no cross-react with recombinant human Cyclophilin A is observed. Detection of mouse and rat Cyclophilin B has not been tested in Immunocytochemistry. | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunocytochemistry, KnockDown | |
549205 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Cyclophilin B, cyclophilin-like protein, CYPB, CYPBEC 5.2.1.8, CYP-S1, MGC2224, OI9MGC14109, peptidyl-prolyl cis-trans isomerase B, peptidylprolyl isomerase B (cyclophilin B), PPIase B, PPIB, Rotamase B, S-cyclophilin, SCYLP | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5479 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction