Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Cyclophilin B Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5410
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5410 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5410 Supplier R&D Systems Supplier No. AF5410
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Cyclophilin B Polyclonal specifically detects Cyclophilin B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry, Knockout Validated.

Specifications

Antigen Cyclophilin B
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL
Gene Accession No. P23284
Gene Alias Cyclophilin B, cyclophilin-like protein, CYPB, CYPBEC 5.2.1.8, CYP-S1, MGC2224, OI9MGC14109, peptidyl-prolyl cis-trans isomerase B, peptidylprolyl isomerase B (cyclophilin B), PPIase B, PPIB, Rotamase B, S-cyclophilin, SCYLP
Gene Symbols PPIB
Host Species Goat
Immunogen E. coli-derived recombinant human Cyclophilin B Asp34-Glu216 Accession # P23284
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5479
Test Specificity Detects human, mouse, and rat Cyclophilin B in Western blots. In Western blots and direct ELISAs, less than 1% cross-reactivity with recombinant human Cyclophilin A is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.