Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Cytochrome c Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB899SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB899SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB899SP Supplier R&D Systems Supplier No. MAB899SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cytochrome c Monoclonal specifically detects Cytochrome c in Human, Mouse, Rat samples. It is validated for Immunoprecipitation.

Specifications

Antigen Cytochrome c
Applications Immunoprecipitation
Classification Monoclonal
Clone 2.7D5
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunoprecipitation 1 ug/10^6 cells
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias CYCHCS, CYCS, cytochrome c, cytochrome c, somatic, THC4
Gene Symbols CYCS
Host Species Mouse
Immunogen Pigeon Cytochrome c
Purification Method Protein A or G purified from ascites
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54205
Test Specificity Detects native human, mouse, and rat Cytochrome c.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.