Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat DGK-epsilon Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5125
This item is not returnable.
View return policy
Description
DGK-epsilon Monoclonal specifically detects DGK-epsilon in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| DGK-epsilon | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| P52429 | |
| DGKE | |
| E. coli-derived recombinant human DGK-epsilon Asn314-Arg435 Accession # P52429 | |
| 100 μg | |
| Primary | |
| Detects human DGK-epsilon in direct ELISAs and human, mouse and rat DGK-epsilon in Western blots. In Western blots, approximately 10-50% cross-reactivity with recombinant human DGK zeta, eta, iota, alpha, theta, gamma, kappa, delta, and beta is observed. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 670914 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DAG kinase epsilon, DAGK5, DAGK6, DGK, DGKE, DGKepsilon, DGK-epsilon, diacylglycerol kinase epsilon, diacylglycerol kinase, epsilon (64kD), diacylglycerol kinase, epsilon 64kDa, Diglyceride kinase epsilon, EC 2.7.1.107 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8526 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction