Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat DGK-theta Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6598SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6598SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6598SP Supplier R&D Systems Supplier No. MAB6598SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

DGK-theta Monoclonal specifically detects DGK-theta in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen DGK-theta
Applications Western Blot
Classification Monoclonal
Clone 652308
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P52824
Gene Alias DAG kinase theta, DAGK, DAGK4EC 2.7.1.107, DAGK7, DGKQ, DGKtheta, DGK-theta, diacylglycerol kinase theta, diacylglycerol kinase, theta (110kD), diacylglycerol kinase, theta 110kDa, Diglyceride kinase theta
Gene Symbols DGKQ
Host Species Mouse
Immunogen E. coli-derived recombinant human DGK-theta Asp696-Lys839 Accession # P52824
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1609
Test Specificity Detects DGK-theta in direct ELISAs and Western blots. In direct ELISAs and Western blolts, no cross-reactivity with recombinant human DGK-alpha , -beta , -gamma , -epsilon , -iota , -kappa , or -zeta is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.