Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Dihydrofolate Reductase/DHFR Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7934SP
This item is not returnable.
View return policy
Description
Dihydrofolate Reductase/DHFR Monoclonal specifically detects Dihydrofolate Reductase/DHFR in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry.Specifications
Dihydrofolate Reductase/DHFR | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
P00374 | |
DHFR | |
E. coli-derived recombinant human Dihydrofolate Reductase/DHFR Met1-Asp187 Accession # P00374 | |
25 μg | |
Primary | |
Detects human Dihydrofolate Reductase/DHFR in ELISA. Detects human, mouse and rat Dihydrofolate Reductase in Western Blots. | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
872442 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DHFR, DHFRP1, dihydrofolate reductase, DYR, EC 1.5.1.3 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1719 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction