Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat DRAK2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6640
This item is not returnable.
View return policy
Description
DRAK2 Monoclonal specifically detects DRAK2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
DRAK2 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
O94768 | |
STK17B | |
E. coli-derived recombinant human DRAK2 Ala87-Glu285 (predicted) Accession # O94768 | |
100 μg | |
Primary | |
Detects human, mouse, and rat DRAK2 in Western blots. In Western blots, less than 10% cross-reactivity with recombinant human (rh) DRAK1, rhDAPK1, rhDAPK2, or rhDAPK3 is observed. | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
685228 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DAP kinase-related apoptosis-inducing protein kinase 2, DRAK2death-associated protein kinase-related 2, EC 2.7.11, EC 2.7.11.1, serine/threonine kinase 17b, serine/threonine kinase 17b (apoptosis-inducing), serine/threonine-protein kinase 17B, STK17B | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
9262 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction