Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat EEA1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB8047
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8047 100 μg
MAB8047SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB8047 Supplier R&D Systems Supplier No. MAB8047
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

EEA1 Monoclonal specifically detects EEA1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen EEA1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 871546
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q15075
Gene Alias early endosome antigen 1, early endosome antigen 1, 162kD, early endosome-associated protein, Endosome-associated protein p162, MST105, MSTP105, ZFYVE2, ZFYVE2MST105, Zinc finger FYVE domain-containing protein 2
Gene Symbols EEA1
Host Species Mouse
Immunogen E. coli-derived recombinant human EEA1 Asn1249-Gln1356 Accession # Q15075
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8411
Test Specificity Detects human EEA1 in direct ELISAs and human, mouse, and rat EEA1 in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.