Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat eIF4E Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB3228SP
This item is not returnable.
View return policy
Description
eIF4E Monoclonal specifically detects eIF4E in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry, Knockout Validated.Specifications
eIF4E | |
Monoclonal | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunocytochemistry 8-25 ug/mL, Knockout Validated | |
P06730 | |
EIF4E | |
E. coli-derived recombinant human eIF4E Met1-Val217 Accession # P06730 | |
25 μg | |
Primary | |
Detects human, mouse and rat eIF4E. | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunocytochemistry, Immunoassay | |
299910 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CBP, eIF4E, eIF-4E, EIF4E1, EIF4EL1, EIF4EL1MGC111573, EIF4F, eIF-4F 25 kDa subunit, eukaryotic translation initiation factor 4E, eukaryotic translation initiation factor 4E-like 1, mRNA cap-binding protein | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1977 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction