Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat EMP/MAEA Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7288
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7288 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7288 Supplier R&D Systems Supplier No. AF7288
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody has been used in 2 publications

EMP/MAEA Polyclonal specifically detects EMP/MAEA in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen EMP/MAEA
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL
Gene Accession No. Q7L5Y9
Gene Alias EMLP, EMPCell proliferation-inducing gene 5 protein, Erythroblast macrophage protein, HLC-10, Human lung cancer oncogene 10 protein, lung cancer-related protein 10, macrophage erythroblast attacher, MAEA, PIG5, proliferation-inducing gene 5
Gene Symbols MAEA
Host Species Sheep
Immunogen E. coli-derived recombinant human EMP/MAEA Met12-Ser66 Accession # Q7L5Y9
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10296
Test Specificity Detects human, mouse and rat EMP/MAEA in Western blots. Detects recombinant human EMP/MAEA in direct ELISAs.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.