Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Endophilin A1/SH3GL2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF8006
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF8006 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF8006 Supplier R&D Systems Supplier No. AF8006
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Endophilin A1/SH3GL2 Polyclonal specifically detects Endophilin A1/SH3GL2 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Endophilin A1/SH3GL2
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q99962
Gene Alias CNSA2, CNSA2FLJ25015, EEN-B1, EEN-B1bA335L15.1 (SH3-domain GRB2-like 2), Endophilin-1, endophilin-A1, FLJ20276, SH3 domain protein 2A, SH3 domain-containing GRB2-like protein 2, SH3D2A, SH3D2AEndophilin A1 BAR domain, SH3-domain GRB2-like 2, SH3GL2, SH3P4, SH3P4endophilin-1
Gene Symbols SH3GL2
Host Species Sheep
Immunogen E. coli-derived recombinant human Endophilin A1/SH3GL2 Ala217-Pro293 Accession # Q99962
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6456
Test Specificity Detects human, mouse, and rat Endophilin A1/SH3GL2 in Western blots.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.