Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Ets-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7284
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7284 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7284 Supplier R&D Systems Supplier No. AF7284
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Ets-1 Polyclonal specifically detects Ets-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Ets-1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL
Gene Accession No. P14921
Gene Alias Avian erythroblastosis virus E26 (v-ets) oncogene homolog-1, ets protein, Ets1, ETS-1, EWSR2, FLJ10768, p54, Protein C-Ets-1, v-ets, v-ets avian erythroblastosis virus E2 oncogene homolog 1, v-ets avian erythroblastosis virus E26 oncogene homolog 1, v-ets erythroblastosis virus E26 oncogene homolog 1 (avian)
Gene Symbols ETS1
Host Species Sheep
Immunogen E. coli-derived recombinant human Ets-1 Glu127-Val230 Accession # P14921
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2113
Test Specificity Detects human, mouse, and rat Ets-1 in Western blots and detects recombinant human Ets-1 in direct ELISAs.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.