Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Ezrin Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7239
This item is not returnable.
View return policy
Description
Ezrin Polyclonal specifically detects Ezrin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Ezrin | |
Polyclonal | |
Unconjugated | |
P15311 | |
EZR | |
E. coli-derived recombinant human Ezrin Lys438-Arg562 Accession # P15311 | |
100 μg | |
Primary | |
Detects human, mouse, and rat Ezrin in Western blots and detects recombinant human Ezrin in direct ELISAs. | |
Human, Mouse, Rat | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.2 ug/mL | |
CVIL, CVL, Cytovillin, cytovillin 2, DKFZp762H157, EZR, ezrin, FLJ26216, p81, VIL2, VIL2MGC1584, Villin 2, villin 2 (ezrin), villin-2 | |
Sheep | |
Affinity Purified | |
RUO | |
7430 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction