Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat FABP1/L-FABP Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7009
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7009 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7009 Supplier R&D Systems Supplier No. AF7009
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

FABP1/L-FABP Polyclonal specifically detects FABP1/L-FABP in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.

Specifications

Antigen FABP1/L-FABP
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Simple Western 1 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. P12710
Gene Alias FABPL, fatty acid binding protein 1, liver, fatty acid-binding protein, liver, LFABP, L-FABP, L-FABPFatty acid-binding protein 1, Liver-type fatty acid-binding protein
Gene Symbols FABP1
Host Species Sheep
Immunogen E. coli-derived recombinant mouse FABP1/L-FABP Met1-Ile127 Accession # P12710
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2168
Test Specificity Detects human, mouse, and rat FABP1/L-FABP in direct ELISAs and Western blots.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.