Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat FABP1/L-FABP Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF1565
This item is not returnable.
View return policy
Description
FABP1/L-FABP Polyclonal specifically detects FABP1/L-FABP in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry.Specifications
FABP1/L-FABP | |
Polyclonal | |
Unconjugated | |
P02692 | |
FABP1 | |
E. coli-derived recombinant rat FABP1/L-FABP Met1-Ile127 Accession # P02692 | |
100 μg | |
Primary | |
Detects rat FABP1/L-FABP in direct ELISAs and Western blots. In Western blots, less than 5% cross-reactivity with recombinant rat FABP2, recombinant human FABP3, recombinant mouse (rm) FABP4, and rmFABP5 is observed. | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
LYOPH | |
Western Blot 0.2-0.5 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL | |
FABPL, fatty acid binding protein 1, liver, fatty acid-binding protein, liver, LFABP, L-FABP, L-FABPFatty acid-binding protein 1, Liver-type fatty acid-binding protein | |
Goat | |
Affinity Purified | |
RUO | |
2168 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction