Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat FABP5/E-FABP Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 9 publications
Supplier: R&D Systems AF1476
This item is not returnable.
View return policy
Description
FABP5/E-FABP Polyclonal specifically detects FABP5/E-FABP in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Knockout Validated.Specifications
| FABP5/E-FABP | |
| Polyclonal | |
| Unconjugated | |
| Q05816 | |
| FABP5 | |
| E. coli-derived recombinant mouse FABP5/E-FABP Met1-Gln135 Accession # Q05816 | |
| 100 μg | |
| Primary | |
| Detects mouse FABP5/E-FABP in direct ELISAs and Western blots. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoassay | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 3-15 ug/mL, Knockout Validated | |
| EFABP, E-FABP, E-FABPPAFABP, fatty acid binding protein 5 (psoriasis-associated), Fatty acid-binding protein 5, PA-FABPepidermal, Psoriasis-associated fatty acid-binding protein homolog | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2171 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction