Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Fc mu R/FAIM3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB9494100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9494100 100 μg
MAB9494SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9494100 Supplier R&D Systems Supplier No. MAB9494100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

FAIM3 Monoclonal specifically detects FAIM3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.

Specifications

Antigen FAIM3
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 992338
Conjugate Unconjugated
Dilution Western Blot 2 μg;/mL, Flow Cytometry 0.25 μg/106 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. O60667
Gene Alias FAIM3, Fas apoptotic inhibitory molecule 3, Fc mu receptor, FCMR, immunoglobulin mu Fc receptor, Regulator of Fas-induced apoptosis Toso, Toso, TOSOIgM Fc receptor
Host Species Mouse
Immunogen Human embryonic kidney cell line HEK293-derived recombinant human Fc mu R/FAIM3, Arg18-Gly251, Accession No. O60667
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 9214.0
Test Specificity Detects human Fc mu R/FAIM3 in direct ELISAs. Detects human, mouse, and rat Fc mu R/FAIM3 in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.