Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Fetuin B Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1725
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1725 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF1725 Supplier R&D Systems Supplier No. AF1725
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Fetuin B Polyclonal specifically detects Fetuin B in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen Fetuin B
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.25 ug/mL, Simple Western 2.5 ug/mL
Gene Accession No. Q9UGM5
Gene Alias FETUB, fetuin-B, Fetuin-like protein IRL685,16G2, Gugu, IRL685
Gene Symbols FETUB
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Fetuin B Met19-Pro382 Accession # Q9UGM5
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26998
Test Specificity Detects human Fetuin B in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) Fetuin A, rhCystatin A, rhCystatin B, rhCystatin C, rhCystatin D, rhCystatin E/M, rhCystatin S, rhCystatin SA, and rhCystatin SN is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.