Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat FKBP12.6 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF4174
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF4174 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF4174 Supplier R&D Systems Supplier No. BAF4174
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

FKBP12.6 Polyclonal specifically detects FKBP12.6 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen FKBP12.6
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 1 ug/mL
Gene Accession No. P68106
Gene Alias 12.6 kDa FK506-binding protein, calstabin 2,12.6 kDa FKBP, EC 5.2.1.8, FK506 binding protein 1B, 12.6 kDa, FK506-binding protein 12.6, FK506-binding protein 1B, FK506-binding protein 1B (12.6 kD), FKBP-12.6, FKBP12.6h-FKBP-12, FKBP1B, FKBP-1B, FKBP1L, FKBP9, Immunophilin FKBP12.6, OTK4, OTK4PPIase FKBP1B, peptidyl-prolyl cis-trans isomerase FKBP1B, PKBP1L, PPIase, Rotamase
Gene Symbols FKBP1B
Host Species Goat
Immunogen E. coli-derived recombinant human FKBP12.6 Met1-Glu108 Accession # P68106
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2281
Test Specificity Detects human, mouse, and rat FKBP12.6 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.