Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat FKBP12/FKBP12.6 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4174100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4174100 100 μg
MAB4174SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB4174100 Supplier R&D Systems Supplier No. MAB4174100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

FKBP12/FKBP12.6 Monoclonal specifically detects FKBP12/FKBP12.6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen FKBP12/FKBP12.6
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 426227
Conjugate Unconjugated
Dilution Western Blot 1 μg;/mL, Immunohistochemistry 5-25 μg;/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P68106
Host Species Rat
Immunogen E. coli-derived recombinant human FKBP12.6, Gly2-Glu108, Accession No. P68106
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Test Specificity Detects human FKBP12.6 in direct ELISAs. Detects human, mouse, and rat FKBP12 and FKBP12.6 in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.