Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat FKBP13 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF4356
This item is not returnable.
View return policy
Description
FKBP13/FKBP2 Polyclonal specifically detects FKBP13/FKBP2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
FKBP13/FKBP2 | |
Polyclonal | |
Unconjugated | |
P26885 | |
FKBP2 | |
E. coli-derived recombinant human FKBP13 Ala22-Leu142 Accession # P26885 | |
100 μg | |
Primary | |
Detects endogenous human, mouse, and rat FKBP13 in Western blots. In Western blots, this antibody does not cross-react with recombinant human FKBP12, FKBP25, FKBP38, FKBP51 or FKBP52. | |
Human, Mouse, Rat | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
EC 5.2.1.8,13 kDa FK506-binding protein, FK506 binding protein 2, 13kDa, FK506-binding protein 2, FK506-binding protein 2 (13kD), FKBP13,13 kDa FKBP, FKBP-13proline isomerase, FKBP2, FKBP-2, Immunophilin FKBP13, peptidyl-prolyl cis-trans isomerase FKBP2, PPIase, PPIase FKBP2, rapamycin-binding protein, Rotamase | |
Sheep | |
Affinity Purified | |
RUO | |
2286 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction