Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat FKBP25 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB3955SP
This item is not returnable.
View return policy
Description
FKBP25 Monoclonal specifically detects FKBP25 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
FKBP25 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q00688 | |
FKBP3 | |
E. coli-derived recombinant human FKBP25 Ala2-Asp224 Accession # Q00688 | |
25 μg | |
Primary | |
Detects endogenous human, mouse, and rat FKBP25 in Western blots. In Western blots, this antibody does not cross-react with recombinant human FKBP12, FKBP12.6, FKBP13, FKBP38, FKBP51, or FKBP52. | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
423325 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 5.2.1.8,25 kDa FK506-binding protein, FK506 binding protein 3, 25kDa, FK506-binding protein 3, FK506-binding protein 3 (25kD), FKBP25,25 kDa FKBP, FKBP3, FKBP-3, Immunophilin FKBP25, peptidyl-prolyl cis-trans isomerase FKBP3, PPIase, PPIase FKBP3, rapamycin binding protein, Rapamycin-selective 25 kDa immunophilin, Rotamase, T-cell | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2287 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction