Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat FKBP38 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB3580
This item is not returnable.
View return policy
Description
FKBP38 Monoclonal specifically detects FKBP38 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
FKBP38 | |
Monoclonal | |
LYOPH | |
Western Blot 0.5 ug/mL | |
Q14318 | |
FKBP8 | |
E. coli-derived recombinant human FKBP38 Met58-Gly382 Accession # Q14318 | |
100 μg | |
Primary | |
Detects endogenous human, mouse and rat FKBP38 in Western blots. | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
391505 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
38 kDa FK506-binding protein, EC 5.2.1.8,38 kDa FKBP, FK506 binding protein 8, 38kDa, FK506-binding protein 8, FK506-binding protein 8 (38kD), FKBP-38, FKBP38PPIase FKBP8, FKBP8, FKBP-8, FKBPr38, hFKBP38, peptidyl-prolyl cis-trans isomerase FKBP8, rotamase | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
23770 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction