Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat FKBP51 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4094
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4094 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4094 Supplier R&D Systems Supplier No. AF4094
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 3 publications

FKBP51/FKBP5 Polyclonal specifically detects FKBP51/FKBP5 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Knockout Validated.

Specifications

Antigen FKBP51/FKBP5
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.3 ug/mL, Simple Western 3 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. Q13451
Gene Alias 51 kDa FKBP, 54 kDa progesterone receptor-associated immunophilin, AIG6,51 kDa FK506-binding protein, Androgen-regulated protein 6, Dit1, FF1 antigen, FK506 binding protein 5, FK506-binding protein 5, FKBP5, FKBP-5, FKBP-51, FKBP51PPIase FKBP5, FKBP54, FKBP54EC 5.2.1.8, HSP90-binding immunophilin, MGC111006, p54, P54,51 kDa FK506-binding protein 5, peptidylprolyl cis-trans isomerase, peptidyl-prolyl cis-trans isomerase FKBP5, PPIase, Ptg-10, Rotamase, T-cell FK506-binding protein
Gene Symbols FKBP5
Host Species Goat
Immunogen E. coli-derived recombinant human FKBP51 Thr2-Val457 Accession # Q13451
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2289
Test Specificity Detects human, mouse, and rat FKBP51. In Western blots, less than 1% cross-reactivity with recombinant human FKBP12, FKBP13, FKBP38, and FKBP52 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.