Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat FKBP52 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4095SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4095SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4095SP Supplier R&D Systems Supplier No. AF4095SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

FKBP52/FKBP4 Polyclonal specifically detects FKBP52/FKBP4 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen FKBP52/FKBP4
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.3 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q02790
Gene Alias 51 kDa FK506-binding protein, 52 kDa FKBP, 59 kDa immunophilin, EC 5.2.1.8, FK506 binding protein 4, 59kDa, FK506-binding protein 4, FK506-binding protein 4 (59kD), FKBP4, FKBP-4, FKBP51, FKBP-52, FKBP52T-cell FK506-binding protein, 59kD, FKBP59, FKBP5952 kDa FK506-binding protein, HBI, HSP binding immunophilin, Hsp56, HSP-binding immunophilin, Immunophilin FKBP52, p52, p59, peptidylprolyl cis-trans isomerase, peptidyl-prolyl cis-trans isomerase FKBP4, PPIase, PPIase FKBP4, Rotamase
Gene Symbols FKBP4
Host Species Goat
Immunogen E. coli-derived recombinant human FKBP52 Met1-Ala459 Accession # Q02790
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2288
Test Specificity Detects endogenous human, mouse and rat FKBP52 in Western blots. In Western blots, this antibody does not cross-react with recombinant human FKBP12, FKBP13, FKBP38 or FKBP51.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.