Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Follistatin-like 4/FSTL4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5245SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5245SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5245SP Supplier R&D Systems Supplier No. AF5245SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Follistatin-like 4/FSTL4 Polyclonal specifically detects Follistatin-like 4/FSTL4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Follistatin-like 4/FSTL4
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q5STE3
Gene Alias Follistatin like 4, follistatin-like 4, Follistatin-like protein 4, follistatin-related protein 4, FSL4, FSTL4, KIAA1061
Gene Symbols FSTL4
Host Species Goat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Follistatin-like 4/FSTL4 Trp23-Val841 Accession # Q5STE3
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23105
Test Specificity Detects recombinant mouse Follistatin-like 4/FSTL4 in direct ELISAs. Detects human, mouse, and rat Follistatin-like 4/FSTL4 in Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) FSTL4 is observed, and less than 5% cross-reactivity with rhFSTL5 and recombinant mouse FSTL1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.