Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat FoxP3 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC8970U100UG
This item is not returnable.
View return policy
Description
FoxP3 Monoclonal specifically detects FoxP3 in Human, Mouse, Rat samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
FoxP3 | |
Monoclonal | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
AIID, AIIDMGC141961, DIETER, forkhead box P3, Forkhead Box Protein P3, FOXP3delta7, immune dysregulation, polyendocrinopathy, enteropathy, X-linked, Immunodeficiency, Polyendocrinopathy, Enteropathy, X-Linked, IPEX, JM2, MGC141961, MGC141963, PIDX, PIDXMGC141963, Scurfin, XPID, XPIDpolyendocrinopathy, enteropathy, X-linked | |
E.coli-derived recombinant human FoxP3 Gln105-Lys200 Accession # Q9BZS1 | |
RUO | |
50943 | |
Human, Mouse, Rat | |
IgG1 |
Flow Cytometry | |
376209R | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9BZS1 | |
Mouse | |
100 μg | |
Primary | |
Detects human FoxP3 in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction