Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat FoxP3 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Catalog No. IC8970R100
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC8970R100 100 Tests
IC8970R025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. IC8970R100 Supplier R&D Systems Supplier No. IC8970R100
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FoxP3 Monoclonal specifically detects FoxP3 in Human, Mouse, Rat samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen FoxP3
Applications Flow Cytometry
Classification Monoclonal
Clone 376209
Conjugate Alexa Fluor 647
Dilution Intracellular Staining by Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q9BZS1
Gene Alias AIID, AIIDMGC141961, DIETER, forkhead box P3, Forkhead Box Protein P3, FOXP3delta7, immune dysregulation, polyendocrinopathy, enteropathy, X-linked, Immunodeficiency, Polyendocrinopathy, Enteropathy, X-Linked, IPEX, JM2, MGC141961, MGC141963, PIDX, PIDXMGC141963, Scurfin, XPID, XPIDpolyendocrinopathy, enteropathy, X-linked
Gene Symbols FOXP3
Host Species Mouse
Immunogen E. coli-derived recombinant human FoxP3 Gln105-Lys200 Accession No. Q9BZS1
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50943
Test Specificity Detects human FoxP3 in direct ELISAs. Detects human, mouse, and rat FoxP3 in flow cytometry.
Target Species Rat, Mouse, Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.