Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat FoxP3 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC3240A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC3240A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC3240A Supplier R&D Systems Supplier No. IC3240A
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

FoxP3 Polyclonal specifically detects FoxP3 in Human, Mouse, Rat samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen FoxP3
Applications Flow Cytometry
Classification Polyclonal
Conjugate APC
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Gene Accession No. Q9BZS1
Gene Alias AIID, AIIDMGC141961, DIETER, forkhead box P3, Forkhead Box Protein P3, FOXP3delta7, immune dysregulation, polyendocrinopathy, enteropathy, X-linked, Immunodeficiency, Polyendocrinopathy, Enteropathy, X-Linked, IPEX, JM2, MGC141961, MGC141963, PIDX, PIDXMGC141963, Scurfin, XPID, XPIDpolyendocrinopathy, enteropathy, X-linked
Gene Symbols FOXP3
Host Species Goat
Immunogen E. coli-derived recombinant human FoxP3 Gln105-Lys200 Accession # Q9BZS1
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50943
Test Specificity Detects human FoxP3 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human FoxD3 is observed.
Target Species Human, Mouse, Rat
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.