Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat FoxP3 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC8970P025
This item is not returnable.
View return policy
Description
FoxP3 Monoclonal specifically detects FoxP3 in Human, Mouse, Rat samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
FoxP3 | |
Monoclonal | |
R-PE | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
AIID, AIIDMGC141961, DIETER, forkhead box P3, Forkhead Box Protein P3, FOXP3delta7, immune dysregulation, polyendocrinopathy, enteropathy, X-linked, Immunodeficiency, Polyendocrinopathy, Enteropathy, X-Linked, IPEX, JM2, MGC141961, MGC141963, PIDX, PIDXMGC141963, Scurfin, XPID, XPIDpolyendocrinopathy, enteropathy, X-linked | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
RUO | |
50943 | |
Human, Mouse, Rat | |
Purified |
Flow Cytometry | |
376209 | |
Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
Q9BZS1 | |
FOXP3 | |
E. coli-derived recombinant human FoxP3 Gln105-Lys200 Accession # Q9BZS1 | |
25 Tests | |
Primary | |
Detects human FoxP3 in direct ELISAs. Detects human, mouse, and rat FoxP3 in flow cytometry. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction