Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat FPRP/PTGFRN Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB10043SP
This item is not returnable.
View return policy
Description
FPRP/PTGFRN Monoclonal specifically detects FPRP/PTGFRN in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
FPRP/PTGFRN | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
CD315, CD9P-1, FPRP, PTGFRN | |
Human embryonic kidney cell, HEK293-derived human FPRP/PTGFRN, Pro27-Pro832, Accession # Q9P2B2 | |
25 μg | |
5738 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG2b |
Western Blot, Immunohistochemistry | |
998175 | |
Western Blot 2 μg/mL, Immunohistochemistry 5-25 μg/mL | |
Q9P2B2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human FPRP/PTGFRN in direct ELISAs. Detects human, mouse, and rat FPRP/PTGFRN in Western blots. | |
Human, Mouse, Rat | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction