Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Frk Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. AF3766 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3766 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3766 Supplier R&D Systems Supplier No. AF3766

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goat Polyclonal Antibody

Frk Polyclonal specifically detects Frk in Human∣Mouse∣Rat samples. It is validated for Immunohistochemistry∣Western Blot.

Specifications

Antigen Frk
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. P42685
Gene Alias EC 2.7.10, fyn-related kinaseProtein-tyrosine kinase 5, Gtk, GTKEC 2.7.10.2, Nuclear tyrosine protein kinase RAK, PTK5, PTK5 protein tyrosine kinase 5, Rak, RAKPTK5, tyrosine-protein kinase FRK
Gene Symbols FRK
Host Species Goat
Immunogen E. coli-derived recombinant human Frk Met1-Glu107 Accession # P42685
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2444
Test Specificity Detects human, mouse, and rat Frk in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Product Type Antibody
Isotype IgG
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.