Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Fyn Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB3574SP
This item is not returnable.
View return policy
Description
Fyn Monoclonal specifically detects Fyn in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry.Specifications
Fyn | |
Monoclonal | |
LYOPH | |
Western Blot 0.5 ug/mL, Immunocytochemistry 8-25 ug/mL | |
P06241 | |
FYN | |
E. coli-derived recombinant human Fyn Met1-Gly83 Accession # P06241 | |
25 μg | |
Primary | |
Detects endogenous human, mouse and rat Fyn in Western blots. | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunocytochemistry | |
361228 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 2.7.10, EC 2.7.10.2, FYN oncogene related to SRC, FGR, YES, MGC45350, OKT3-induced calcium influx regulator, p59-Fyn, protein-tyrosine kinase fyn, Proto-oncogene c-Fyn, Proto-oncogene Syn, proto-oncogene tyrosine-protein kinase fyn, SLK, SLKc-syn protooncogene, src/yes-related novel, Src-like kinase, SYN, tyrosine kinase p59fyn(T), tyrosine-protein kinase Fyn | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2534 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction