Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat GABA-A R alpha 4 N-Terminus Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Supplier: R&D Systems PPS026
This item is not returnable.
View return policy
Description
GABA-AR alpha 4 Polyclonal specifically detects GABA-AR alpha 4 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
GABA-AR alpha 4 | |
Polyclonal | |
Western Blot 1:1000 dilution | |
GABRA4 | |
Peptide from GABA A R alpha 4 subunit, N-Terminus conjugated to keyhole limpet hemocyanin (KLH) | |
100 μL | |
Primary | |
Human, mouse, rat, and bovine ∼64 kDa GABA A R alpha 4 subunit, N-Terminus | |
For long-term storage, < -20degrees C is recommended. Product is stable at < -20degrees C for at least 1 year. |
Western Blot | |
Unconjugated | |
GABA(A) receptor subunit alpha-4, GABAAR alpha 4, GABAARa4, GABRA4, gamma-aminobutyric acid (GABA) A receptor, alpha 4, gamma-aminobutyric acid receptor subunit alpha-4 | |
Rabbit | |
Affinity Purified | |
RUO | |
2557 | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction