Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat GABAB R2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB10456SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10456SP 25 μg
MAB10456100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB10456SP Supplier R&D Systems Supplier No. MAB10456SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

GABA-B R2 Monoclonal antibody specifically detects GABA-B R2 in Human, Mouse, Rat samples. It is validated for Western Blot, Western Blot, Immunohistochemistry

Specifications

Antigen GABA-B R2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 2605C
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. O88871
Gene Alias G protein-coupled receptor 51, GABABR2, GABA-BR2, GABA-B-R2, GABABR2GABA-B receptor 2, GABBR2, gamma-aminobutyric acid (GABA) B receptor, 2, gamma-aminobutyric acid type B receptor subunit 2, gb2, GPR51, GPR51GABA-B receptor, R2 subunit, GPRC3B, GPRC3BFLJ36928, G-protein coupled receptor 51, HG20HRIHFB2099
Host Species Rabbit
Immunogen Mouse myeloma cell line NS0-derived recombinant rat GABAB R2, Trp41-Ser482, Accession # O88871
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9568
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.