Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat GAPDH Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. 2275PC100
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2275PC100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 2275PC100 Supplier R&D Systems Supplier No. 2275PC100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 62 publications

GAPDH Polyclonal specifically detects GAPDH in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunoprecipitation.

Specifications

Antigen GAPDH
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Simple Western 1 ug/mL, Immunoprecipitation 1:1000-1:5000 dilution
Gene Alias aging-associated gene 9 protein, EC 1.2.1, EC 1.2.1.12, EC 2.6.99.-, G3PD, G3PDH, GAPD, glyceraldehyde 3-phosphate dehydrogenase, glyceraldehyde-3-phosphate dehydrogenase, MGC88685, Peptidyl-cysteine S-nitrosylase GAPDH
Gene Symbols GAPDH
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a portion of the human glyceraldehyde-3-phosphate dehydrogenase (GAPDH) sequence
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2597
Test Specificity The antibody detects human and mouse G3PDH (other species not tested).
Target Species Human, Mouse, Rat
Content And Storage This antibody can be stored at -20degreesC or -80degreesC. Avoid repeated freeze-thawing by aliquoting into smaller portions.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.