Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat GAPDH Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF5718SP
This item is not returnable.
View return policy
Description
GAPDH Polyclonal specifically detects GAPDH in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry.Specifications
GAPDH | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
aging-associated gene 9 protein, EC 1.2.1, EC 1.2.1.12, EC 2.6.99.-, G3PD, G3PDH, GAPD, glyceraldehyde 3-phosphate dehydrogenase, glyceraldehyde-3-phosphate dehydrogenase, MGC88685, Peptidyl-cysteine S-nitrosylase GAPDH | |
Goat | |
Affinity Purified | |
RUO | |
2597 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-25 ug/mL | |
P04406 | |
GAPDH | |
E. coli-derived recombinant human GAPDH/G3PDH Met1-Ala150 Accession # P04406 | |
25 μg | |
Primary | |
Detects human, mouse, and rat GAPDH/G3PDH in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human GAPDH-2 is observed. | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction