Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat GATE-16 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $502.50
Specifications
Antigen | GATE-16/GABARAPL2 |
---|---|
Clone | 853746 |
Dilution | Western Blot 2 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
GATE-16/GABARAPL2 Monoclonal specifically detects GATE-16/GABARAPL2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
GATE-16/GABARAPL2 | |
Western Blot 2 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Apg8p2, ATG8, ATG8C, FLC3A, GABARAPL2, gamma-aminobutyric acid receptor-associated protein-like 2, Ganglioside expression factor 2, GATE16, GATE-16, GEF2, GEF-2, GEF2GEF-2, General Protein Transport Factor P16, Golgi-Associated ATPase Enhancer Of 16 KDa, MAP1 light chain 3 related protein, MAP1 light chain 3-related protein | |
GABARAPL2 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
853746 | |
Western Blot | |
Unconjugated | |
Rat | |
Human, Mouse, Rat | |
P60520 | |
11345 | |
E.coli-derived recombinant human GATE16 Accession # P60520 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human, mouse, and rat GATE-16 in Western blot. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title