Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Gelsolin/GSN Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB8170
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8170 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB8170 Supplier R&D Systems Supplier No. MAB8170
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Gelsolin/GSN Monoclonal specifically detects Gelsolin/GSN in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.

Specifications

Antigen Gelsolin/GSN
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 893205
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunohistochemistry 8-25 ug/mL
Gene Accession No. P06396
Gene Alias Actin-depolymerizing factor, ADF, AGEL, Brevin, DKFZp313L0718, gelsolin, gelsolin (amyloidosis, Finnish type), GSN
Gene Symbols GSN
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human Gelsolin/GSN Met1-Ala782 Accession # P06396
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 2934
Test Specificity Detects human Gelsolin/GSN in ELISAs. Detects human, mouse and rat Gelsolin/GSN in Western Blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.