Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat GIT1 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC8508V100UG
This item is not returnable.
View return policy
Description
GIT1 Monoclonal specifically detects GIT1 in Human, Mouse, Rat samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
GIT1 | |
Monoclonal | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
ARF GAP GIT1, ARF GTPase-activating protein GIT1, CAT1, CAT-1, Cool-associated and tyrosine-phosphorylated protein 1, G protein-coupled receptor kinase interacting ArfGAP 1, G protein-coupled receptor kinase interactor 1, G protein-coupled receptor kinase-interactor 1, GRK-interacting protein 1 | |
E. coli-derived recombinant human GIT1 Ser485-Asp636 Accession # Q9Y2X7 | |
RUO | |
28964 | |
Human, Mouse, Rat | |
IgG2b |
Flow Cytometry | |
924640 | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9Y2X7 | |
Mouse | |
100 μg | |
Primary | |
Detects human GIT1 in direct ELISAs and human, mouse, and rat GIT1 in Western blots. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction