Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Glutaredoxin 3/GLRX3 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7560
This item is not returnable.
View return policy
Description
Glutaredoxin 3/GLRX3 Polyclonal specifically detects Glutaredoxin 3/GLRX3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Glutaredoxin 3/GLRX3 | |
Polyclonal | |
Unconjugated | |
O76003 | |
GLRX3 | |
E. coli-derived recombinant human Glutaredoxin 3/GLRX3 Asn126-Lys294 Accession # O76003 | |
100 μg | |
Primary | |
Detects human, mouse and rat Glutaredoxin 3/GLRX3 in Western blots. | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
FLJ11864, GLRX3, GLRX4, glutaredoxin 3, glutaredoxin 4, glutaredoxin 5, GRX3, GRX4, PICOT, PICOTbA500G10.4, PKC-interacting cousin of thioredoxin, PKCq-interacting protein, thioredoxin-like 2, TXNL2, TXNL3 | |
Sheep | |
Affinity Purified | |
RUO | |
10539 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction