Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Glutathione Peroxidase 1/GPX1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 15 publications
Supplier: R&D Systems AF3798SP
This item is not returnable.
View return policy
Description
Glutathione Peroxidase 1/GPX1 Polyclonal specifically detects Glutathione Peroxidase 1/GPX1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Knockout Validated.Specifications
Glutathione Peroxidase 1/GPX1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Cellular glutathione peroxidase, CGPx, EC 1.11.1, EC 1.11.1.9, glutathione peroxidase 1, GPx-1, GPX1, GSHPx-1, GSHPX1, MGC14399, MGC88245 | |
Goat | |
Affinity Purified | |
RUO | |
2876 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry, KnockDown | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL, Knockout Validated | |
P07203 | |
GPX1 | |
E. coli-derived recombinant human Glutathione Peroxidase 1 Gly48-Ala201 Accession # P07203 | |
25 μg | |
Primary | |
Detects endogenous human, mouse, and rat GPX1. In Western blots, this antibody shows no cross-reactivity with recombinant human GPX2 or GPX3. | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction