Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Glutathione Peroxidase 1/GPX1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3798
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3798 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3798 Supplier R&D Systems Supplier No. AF3798
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 15 publications

Glutathione Peroxidase 1/GPX1 Polyclonal specifically detects Glutathione Peroxidase 1/GPX1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Knockout Validated.

Specifications

Antigen Glutathione Peroxidase 1/GPX1
Applications Western Blot, Immunohistochemistry, KnockDown
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL, Knockout Validated
Gene Accession No. P07203
Gene Alias Cellular glutathione peroxidase, CGPx, EC 1.11.1, EC 1.11.1.9, glutathione peroxidase 1, GPx-1, GPX1, GSHPx-1, GSHPX1, MGC14399, MGC88245
Gene Symbols GPX1
Host Species Goat
Immunogen E. coli-derived recombinant human Glutathione Peroxidase 1 Gly48-Ala201 Accession # P07203
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2876
Test Specificity Detects endogenous human, mouse, and rat GPX1. In Western blots, this antibody shows no cross-reactivity with recombinant human GPX2 or GPX3.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.