Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Glutathione Peroxidase 3/GPX3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4199
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4199 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4199 Supplier R&D Systems Supplier No. AF4199
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 4 publications

Glutathione Peroxidase 3/GPX3 Polyclonal specifically detects Glutathione Peroxidase 3/GPX3 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.

Specifications

Antigen Glutathione Peroxidase 3/GPX3
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Simple Western 25 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. P46412
Gene Alias EC 1.11.1, EC 1.11.1.9, Extracellular glutathione peroxidase, glutathione peroxidase 3, glutathione peroxidase 3 (plasma), GPx-3, GPX3, GPx-P, GPXP, GSHPx-3, GSHPx-P, Plasma glutathione peroxidase
Gene Symbols GPX3
Host Species Goat
Immunogen E. coli-derived recombinant mouse Glutathione Peroxidase 3/GPX3 Gly74-Lys226 Accession # P46412
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2878
Test Specificity Detects human, mouse, and rat Glutathione Peroxidase 3/GPX3 in Western blots and detects recombinant human and recombinant mouse Glutathione Peroxidase 3/GPX3 in direct ELISAs. In direct ELISAs, approximately 15% cross-reactivity with recombinant human (rh) GPX5 and recombinant mouse (rm) GPX1 is observed, and less than 4% cross-reactivity with rhGPX4, rhGPX6, and rmGPX2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.