Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Glyoxalase I Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF4959SP
This item is not returnable.
View return policy
Description
Glyoxalase I Polyclonal specifically detects Glyoxalase I in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western.Specifications
Glyoxalase I | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Aldoketomutase, EC 4.4.1.5, GLO1, GLOD1, Glx I, GLYI, glyoxalase domain containing 1, glyoxalase Ialdoketomutase, Ketone-aldehyde mutase, lactoyl glutathione lyase, lactoylglutathione lyase, methylglyoxalase, S-D-lactoylglutathione methylglyoxal lyase | |
Goat | |
Affinity Purified | |
RUO | |
2739 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 50 ug/mL | |
Q04760 | |
GLO1 | |
E. coli-derived recombinant human Glyoxalase I Ala2-Met184 Accession # Q04760 | |
25 μg | |
Primary | |
Detects human, mouse and rat Glyoxalase I in Western blots. | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction