Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Glyoxalase II Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5944
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5944 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5944 Supplier R&D Systems Supplier No. AF5944
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Glyoxalase II/HAGH Polyclonal specifically detects Glyoxalase II/HAGH in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Glyoxalase II/HAGH
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q16775
Gene Alias EC 3.1.2.6, GLO2hydroxyacyl glutathione hydrolase, Glx II, GLX2, GLXII, Glyoxalase II, HAGH, HAGH1GLX2, hydroxyacylglutathione hydrolase, hydroxyacylglutathione hydrolase, mitochondrial, hydroxyacylglutathione hydroxylase
Gene Symbols HAGH
Host Species Goat
Immunogen E. coli-derived recombinant human Glyoxalase II Met49-Ile254 Accession # Q16775
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3029
Test Specificity Detects human, mouse, and rat Glyoxalase II in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human Glyoxalase I is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.