Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat gp96/HSP90B1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7606
This item is not returnable.
View return policy
Description
gp96/HSP90B1/GRP94 Polyclonal specifically detects gp96/HSP90B1/GRP94 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Knockout Validated.Specifications
gp96/HSP90B1/GRP94 | |
Polyclonal | |
Unconjugated | |
P14625 | |
HSP90B1 | |
E. coli-derived recombinant human gp96/HSP90B1 Arg503-Arg660 Accession # P14625 | |
100 μg | |
Primary | |
Detects human, mouse, and rat gp96/HSP90B1 in direct ELISAs and Western blots. | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry, KnockDown | |
LYOPH | |
Western Blot 0.5 ug/mL, Immunohistochemistry 1-15 ug/mL, Knockout Validated | |
94 kDa glucose-regulated protein, ECGP, Endoplasmin, endothelial cell (HBMEC) glycoprotein, glucose regulated protein, 94 kDa, GP96ECGP, Grp94, GRP-94, GRP94gp96 homolog, Heat shock protein 90 kDa beta member 1, heat shock protein 90kDa beta (Grp94), member 1, HSP90B1, TRA1endoplasmin, tumor rejection antigen (gp96) 1, Tumor rejection antigen 1, Tumor rejection antigen-1 (gp96) | |
Sheep | |
Affinity Purified | |
RUO | |
7184 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction